2-Ethoxy-5-fluorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

22829-1G 22829-25G 22829-500MG 22829-5G
76898-916EA 155.96 USD
76898-916 76898-918 76898-920 76898-922
2-Ethoxy-5-fluorophenol ≥97% research grade
2-Ethoxy-5-fluorophenol
Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18968886
CAS Number: 376600-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR