6-Chloro-5-fluoropyridine-3-carbaldehyde ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
22764-1G
22764-25G
22764-500MG
22764-5G
76898-892EA
153.84
USD
76898-892
76898-894
76898-896
76898-898
6-Chloro-5-fluoropyridine-3-carbaldehyde ≥97% research grade
6-Chloro-5-fluoropyridine-3-carbaldehyde
Formula:
C₆H₃ClFNO Storage Temperature: Ambient |
MDL Number:
MFCD13189060 CAS Number: 950691-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...