2-(2-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester
22717-25G
22717-5G
76898-886EA
453.01
USD
76898-886
76898-888
2-(2-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(2-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula:
C₁₃H₁₅BClF₃O₂ MW: 306.52 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16996251 CAS Number: 1146214-95-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...