2-Methyl-3-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methylpyridine-3-boronic acid pinacol ester, 2-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

22678-25G 22678-5G
76898-880EA 289.63 USD
76898-880 76898-882
2-Methyl-3-pyridineboronic acid pinacol ester ≥97%
2-Methyl-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₂
MW: 219.0878 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669594
CAS Number: 1012084-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR