2,6-Dimethyl-4-isopropoxyphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

22664-1G 22664-25G 22664-500MG 22664-5G
76898-864EA 260.99 USD
76898-864 76898-866 76898-868 76898-870
2,6-Dimethyl-4-isopropoxyphenylboronic acid ≥97% research grade
2,6-Dimethyl-4-isopropoxyphenylboronic acid
Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD18384078
CAS Number: 1192107-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR