2-Bromo-1,3-dimethyl-5-(1-methylethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

22660-1G 22660-25G 22660-500MG 22660-5G
76898-850EA 127.31 USD
76898-850 76898-852 76898-854 76898-856
2-Bromo-1,3-dimethyl-5-(1-methylethoxy)benzene ≥95%
2-Bromo-1,3-dimethyl-5-(1-methylethoxy)benzene
Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18917820
CAS Number: 91345-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR