Methyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methyl-4-methoxycarbonylphenylboronic acid, pinacol ester

22617-25G 22617-5G
76898-842EA 262.05 USD
76898-842 76898-844
Methyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate ≥97%
Methyl 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Formula: C₁₅H₂₁BO₄
MW: 276.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669595
CAS Number: 473596-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR