2-Fluoro-1,3-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

22601-1G 22601-25G 22601-5G
76898-828EA 76.39 USD
76898-828 76898-830 76898-832
2-Fluoro-1,3-dimethoxybenzene ≥97%
2-Fluoro-1,3-dimethoxybenzene
Formula: C₈H₉FO₂
MW: 156.154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547873
CAS Number: 195136-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR