5-(Benzyloxy)-2-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

22598-1G 22598-25G 22598-500MG 22598-5G
76898-816EA 242.95 USD
76898-816 76898-818 76898-820 76898-822
5-(Benzyloxy)-2-chloropyridine ≥97%
5-(Benzyloxy)-2-chloropyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11976457
CAS Number: 84611-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR