2-Chloro-5-(methoxymethoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

22594-1G 22594-25G 22594-5G
76898-802EA 117.76 USD
76898-802 76898-804 76898-806
2-Chloro-5-(methoxymethoxy)pyridine ≥97%
2-Chloro-5-(methoxymethoxy)pyridine

Precaución: for Research Use Only

Formula: C₇H₈ClNO₂
MW: 173.597 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20278147
CAS Number: 877133-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR