2-Fluoro-3-iodobenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

22584-1G 22584-25G 22584-500MG 22584-5G
76898-782EA 224.92 USD
76898-782 76898-784 76898-786 76898-788
2-Fluoro-3-iodobenzyl alcohol ≥97% research grade
2-Fluoro-3-iodobenzyl alcohol
Formula: C₇H₆FIO
Storage Temperature: Refrigerator
MDL Number: MFCD11110181
CAS Number: 307975-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR