2-Fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22582-1G 22582-25G 22582-5G
76898-770EA 112.46 USD
76898-770 76898-772 76898-774
2-Fluoro-3-formylbenzoic acid ≥97%
2-Fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅FO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20526383
CAS Number: 1289043-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR