2-Bromo-5-methoxy-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

22545-1G 22545-25G 22545-500MG 22545-5G
76898-758EA 236.59 USD
76898-758 76898-760 76898-762 76898-764
2-Bromo-5-methoxy-3-methylpyridine ≥97%
2-Bromo-5-methoxy-3-methylpyridine
Formula: C₇H₈BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD18255401
CAS Number: 1256809-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR