2-Fluoro-5-hydroxy-3-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-hydroxy-3-picoline, 6-Fluoro-5-methylpyridin-3-ol

22543-25G 22543-5G
76898-746EA 386.17 USD
76898-746 76898-748
2-Fluoro-5-hydroxy-3-methylpyridine ≥97% research grade
2-Fluoro-5-hydroxy-3-methylpyridine
Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD09839279
CAS Number: 186593-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR