2,5-Difluoro-4-(phenylmethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22479-1G 22479-25G 22479-500MG 22479-5G
76898-706EA 290.69 USD
76898-706 76898-708 76898-710 76898-712
2,5-Difluoro-4-(phenylmethoxy)benzaldehyde ≥97%
2,5-Difluoro-4-(phenylmethoxy)benzaldehyde
Formula: C₁₄H₁₀F₂O₂
MW: 248.225 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28503327
CAS Number: 918524-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR