2-Bromo-4-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

22389-1G 22389-25G 22389-500MG 22389-5G
76898-652EA 256.74 USD
76898-652 76898-654 76898-656 76898-658
2-Bromo-4-methylbenzamide ≥97%
2-Bromo-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD18392004
CAS Number: 131002-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR