2-Methoxypyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

22117-25G 22117-5G
76898-596EA 310.85 USD
76898-596 76898-598
2-Methoxypyridin-3-ol ≥97%
2-Methoxypyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MW: 125.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00457990
CAS Number: 13472-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR