2-Formyl-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22020-1G 22020-500MG 22020-5G
76898-572EA 500.75 USD
76898-572 76898-574 76898-576
2-Formyl-4-methylbenzoic acid ≥97%
2-Formyl-4-methylbenzoic acid
Formula: C₉H₈O₃
MW: 164.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18451214
CAS Number: 61099-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR