2-Fluoro-3-methylpyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

21995-1G 21995-500MG 21995-5G
76898-562EA 498.63 USD
76898-562 76898-564 76898-566
2-Fluoro-3-methylpyridine-4-carboxylic acid ≥97%
2-Fluoro-3-methylpyridine-4-carboxylic acid
Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611272
CAS Number: 885589-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR