2,6-Dibromo-3-hydroxypyridine ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2,6-Dibromopyridin-3-ol
21972-25G
21972-5G
76898-540EA
310.85
USD
76898-540
76898-542
2,6-Dibromo-3-hydroxypyridine ≥95%
2,6-Dibromo-3-hydroxypyridine
Precaución: for Research Use Only
Formula:
C₅H₃Br₂NO Storage Temperature: Ambient |
MDL Number:
MFCD01318479 CAS Number: 6602-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...