2,5-Dibromo-3-(hydroxymethyl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

21828-1G 21828-25G 21828-500MG 21828-5G
76898-508EA 153.84 USD
76898-508 76898-510 76898-512 76898-514
2,5-Dibromo-3-(hydroxymethyl)pyridine ≥97% research grade
2,5-Dibromo-3-(hydroxymethyl)pyridine
Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16606959
CAS Number: 1227490-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR