2-Fluoro-6-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

21758-25G 21758-5G
76898-468EA 325.7 USD
76898-468 76898-470
2-Fluoro-6-methoxybenzyl alcohol ≥97%
2-Fluoro-6-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD00671766
CAS Number: 253668-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR