(6-Chloro-5-methylpyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-hydroxymethyl-3-methylpyridine

21589-1G 21589-25G 21589-500MG 21589-5G
76898-434EA 120.95 USD
76898-434 76898-436 76898-438 76898-440
(6-Chloro-5-methylpyridin-3-yl)methanol ≥97%
(6-Chloro-5-methylpyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MW: 157.598 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09261083
CAS Number: 887707-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR