2-Bromo-1-methyl-4-propoxybenzene ≥97% research grade

Supplier: AOBChem USA

Warning

21459-1G 21459-25G 21459-500MG 21459-5G
76898-390EA 231.28 USD
76898-390 76898-392 76898-394 76898-396
2-Bromo-1-methyl-4-propoxybenzene ≥97% research grade
2-Bromo-1-methyl-4-propoxybenzene
Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19105376
CAS Number: 1026796-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR