2,5-Dichloro-4-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

21408-25G 21408-5G
76898-380EA 616.39 USD
76898-380 76898-382
2,5-Dichloro-4-methoxypyridine ≥97%
2,5-Dichloro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185541
CAS Number: 1122090-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR