2-Fluoro-3-methyl-5-pyridineboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-methylpyridine-5-boronic acid, pinacol ester, 2-Fluoro-3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

21328-1G 21328-25G 21328-5G
76898-370EA 109.28 USD
76898-370 76898-372 76898-374
2-Fluoro-3-methyl-5-pyridineboronic acid pinacol ester ≥97%
2-Fluoro-3-methyl-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
MW: 237.800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11855990
CAS Number: 1150561-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR