2-Bromo-6-(methoxymethyl)pyridine 97%

Supplier: AOBChem USA

Warning

21268-1G 21268-500MG 21268-5G 21268-25G
76898-358EA 558.04 USD
76898-358 76898-360 76898-362 77657-992
2-Bromo-6-(methoxymethyl)pyridine 97%
2-Bromo-6-(methoxymethyl)pyridine
Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20482703
CAS Number: 112575-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR