(4-Ethoxy-2,5-difluorophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Difluoro-4-ethoxyphenylboronic acid

21162-1G 21162-25G 21162-500MG 21162-5G
76898-310EA 126.25 USD
76898-310 76898-312 76898-314 76898-316
(4-Ethoxy-2,5-difluorophenyl)boronic acid ≥97% research grade
(4-Ethoxy-2,5-difluorophenyl)boronic acid
Formula: C₈H₉BF₂O₃
MW: 201.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026701
CAS Number: 1452575-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR