2-Fluoro-6-iodophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

20988-1G 20988-25G 20988-5G
76898-260EA 62.6 USD
76898-260 76898-262 76898-264
2-Fluoro-6-iodophenylboronic acid pinacol ester ≥97%
2-Fluoro-6-iodophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BFIO₂
Storage Temperature: Ambient
MDL Number: MFCD22493821
CAS Number: 1010839-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR