2-Bromo-4-isopropylphenol ≥97%

Supplier: AOBChem USA

Warning

20959-1G 20959-25G 20959-500MG 20959-5G
76898-248EA 185.66 USD
76898-248 76898-250 76898-252 76898-254
2-Bromo-4-isopropylphenol ≥97%
2-Bromo-4-isopropylphenol
Formula: C₉H₁₁BrO
Storage Temperature: Refrigerator
MDL Number: MFCD08700373
CAS Number: 19432-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR