2-Chloro-6-fluoro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

20922-25G 20922-5G
76898-234EA 310.85 USD
76898-234 76898-236
2-Chloro-6-fluoro-3-methylbenzyl alcohol ≥97%
2-Chloro-6-fluoro-3-methylbenzyl alcohol
Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD01631362
CAS Number: 261762-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR