2-Chloro-6-fluoro-m-cresol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-6-fluoro-3-methylphenol, 2-Choro-6-fluoro-3-methylphenol

20920-25G 20920-5G
76898-226EA 212.18 USD
76898-226 76898-228
2-Chloro-6-fluoro-m-cresol ≥97%
2-Chloro-6-fluoro-m-cresol

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631366
CAS Number: 261762-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR