2-Fluoro-5-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-hydroxyanisole

20852-1G 20852-25G 20852-5G
76898-204EA 82.76 USD
76898-204 76898-206 76898-208
2-Fluoro-5-methoxyphenol ≥97%
2-Fluoro-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂
MW: 142.128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16998752
CAS Number: 117902-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR