3-(2,2-Dimethylpropionylamino)isonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(2,2-Dimethylpropanamido)pyridine-4-barboxylic acid

20821-1G 20821-25G 20821-5G
76898-170EA 106.09 USD
76898-170 76898-172 76898-174
3-(2,2-Dimethylpropionylamino)isonicotinic acid ≥97%
3-(2,2-Dimethylpropionylamino)isonicotinic acid
Formula: C₁₁H₁₄N₂O₃
MW: 222.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06659008
CAS Number: 86847-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR