6-Methoxy-m-toluic acid ≥97%

Supplier: AOBChem USA

Warning

20704-25G 20704-5G
76898-154EA 240.83 USD
76898-154 76898-156
6-Methoxy-m-toluic acid ≥97%
6-Methoxy-m-toluic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00045849
CAS Number: 25045-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR