2-Methoxy-5-(morpholinomethyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20700-1G 20700-25G 20700-500MG 20700-5G
76898-140EA 290.69 USD
76898-140 76898-142 76898-144 76898-146
2-Methoxy-5-(morpholinomethyl)phenylboronic acid ≥97% research grade
2-Methoxy-5-(morpholinomethyl)phenylboronic acid
Formula: C₁₂H₁₈BNO₄
Storage Temperature: Ambient
MDL Number: MFCD16198063
CAS Number: 1334321-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR