2-Fluoro-5-hydroxy-3-methoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20640-1G 20640-25G 20640-500MG 20640-5G
76898-112EA 264.17 USD
76898-112 76898-114 76898-116 76898-118
2-Fluoro-5-hydroxy-3-methoxypyridine ≥97% research grade
2-Fluoro-5-hydroxy-3-methoxypyridine
Formula: C₆H₆FNO₂
MW: 143.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611061
CAS Number: 1227511-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR