2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Borono-4-fluorobenzohydrazide, 2-Fluoro-5-(hydrazinecarbonyl)phenylboronic acid

20592-1G 20592-25G 20592-500MG 20592-5G
76898-088EA 177.18 USD
76898-088 76898-090 76898-092 76898-094
2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid ≥95%
2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08436072
CAS Number: 874289-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR