2-Methoxypyrimidine-5-carboxylic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20563-1G 20563-25G 20563-500MG 20563-5G
76898-046EA 138.98 USD
76898-046 76898-048 76898-050 76898-052
2-Methoxypyrimidine-5-carboxylic acid ≥97% research grade
2-Methoxypyrimidine-5-carboxylic acid
Formula: C₆H₆N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08144700
CAS Number: 344325-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR