2-Bromo-4-chloro-3-fluoropyridine 97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-chloro-3-fluoro-pyridine

20524-1G 20524-500MG 20524-5G 20524-25G
76898-024EA 571.83 USD
76898-024 76898-026 76898-028 77654-128
2-Bromo-4-chloro-3-fluoropyridine 97%
2-Bromo-4-chloro-3-fluoropyridine
Formula: C₅H₂BrClFN
MW: 210.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257572
CAS Number: 1155847-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR