2-Bromo-6-fluoro-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

20511-1G 20511-25G 20511-5G
76898-012EA 88.06 USD
76898-012 76898-014 76898-016
2-Bromo-6-fluoro-3-methylphenol ≥97%
2-Bromo-6-fluoro-3-methylphenol
Formula: C₇H₆BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD16295100
CAS Number: 1226808-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR