2-Chloro-5-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

20501-25G 20501-5G
76898-000EA 209 USD
76898-000 76898-002
2-Chloro-5-methoxybenzaldehyde ≥97%
2-Chloro-5-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845925
CAS Number: 13719-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR