2-Fluoro-5-methoxypyridine-4-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

20461-1G 20461-25G 20461-500MG 20461-5G
76897-980EA 379.81 USD
76897-980 76897-982 76897-984 76897-986
2-Fluoro-5-methoxypyridine-4-boronic acid ≥97% research grade
2-Fluoro-5-methoxypyridine-4-boronic acid
Formula: C₆H₇BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD16611245
CAS Number: 1451392-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR