2-Fluoro-5-methoxypyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

20457-1G 20457-25G 20457-500MG 20457-5G
76897-964EA 290.69 USD
76897-964 76897-966 76897-968 76897-970
2-Fluoro-5-methoxypyridine-3-carboxylic acid ≥97%
2-Fluoro-5-methoxypyridine-3-carboxylic acid
Formula: C₇H₆FNO₃
MW: 171.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611226
CAS Number: 1215868-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR