2,5-Dimethoxy-6-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

20451-1G 20451-25G 20451-500MG 20451-5G
76897-948EA 194.15 USD
76897-948 76897-950 76897-952 76897-954
2,5-Dimethoxy-6-fluorophenylboronic acid ≥97%
2,5-Dimethoxy-6-fluorophenylboronic acid
Formula: C₈H₁₀BFO₄
Storage Temperature: Refrigerator
MDL Number: MFCD28142580
CAS Number: 2121511-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR