2,5-Dibromo-4-(hydroxymethyl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20348-1G 20348-25G 20348-500MG 20348-5G
76897-902EA 144.29 USD
76897-902 76897-904 76897-906 76897-908
2,5-Dibromo-4-(hydroxymethyl)pyridine ≥97% research grade
2,5-Dibromo-4-(hydroxymethyl)pyridine
Formula: C₆H₅Br₂NO
MW: 266.918 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847303
CAS Number: 1227563-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR