2-Fluoro-4-isopropoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

20311-1G 20311-25G 20311-500MG 20311-5G
76897-888EA 263.11 USD
76897-888 76897-890 76897-892 76897-894
2-Fluoro-4-isopropoxybenzaldehyde ≥95%
2-Fluoro-4-isopropoxybenzaldehyde
Formula: C₁₀H₁₁FO₂
MW: 182.192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17011999
CAS Number: 1242070-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR