2-Fluoro-5-(methylthio)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
20279-1G
20279-25G
20279-500MG
20279-5G
76897-848EA
204.76
USD
76897-848
76897-850
76897-852
76897-854
2-Fluoro-5-(methylthio)benzoic acid ≥97%
2-Fluoro-5-(methylthio)benzoic acid
Formula:
C₈H₇FO₂S Storage Temperature: Ambient |
MDL Number:
MFCD16036292 CAS Number: 57318-98-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...