2,6-Difluoro-4-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Difluoro-4-formylphenylboronic acid, pinacol ester, 3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

20122-25G 20122-5G
76897-788EA 559.1 USD
76897-788 76897-790
2,6-Difluoro-4-formylphenylboronic acid pinacol ester ≥97%
2,6-Difluoro-4-formylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₂O₃
MW: 268.600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276848
CAS Number: 870717-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR