2-Chloro-3,4-difluorophenylboronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

19979-1G 19979-500MG 19979-5G
76897-766EA 772.34 USD
76897-766 76897-768 76897-770
2-Chloro-3,4-difluorophenylboronic acid ≥95% research grade
2-Chloro-3,4-difluorophenylboronic acid
Formula: C₆H₄BClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18415642
CAS Number: 2121511-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR